3-Chloropropyl isocyanate, 96%, Thermo Scientific™
3-Chloropropyl isocyanate, 96%, Thermo Scientific™
3-Chloropropyl isocyanate, 96%, Thermo Scientific™
3-Chloropropyl isocyanate, 96%, Thermo Scientific™
3-Chloropropyl isocyanate, 96%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA314590050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Chloropropyl isocyanate
Name Note96%
CAS13010-19-0
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger