1,4-Diisocyanatobutane, 97%
1,4-Diisocyanatobutane, 97%
1,4-Diisocyanatobutane, 97%
1,4-Diisocyanatobutane, 97%
Thermo Scientific Chemicals
1,4-Diisocyanatobutane, 97%
CAS: 4538-37-8 | C6H8N2O2 | 140.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA314380250
Price (MYR)
7,844.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
7,844.00
EA
Add to cart
Specifications
Chemical Name or Material1, 4-Diisocyanatobutane
CAS4538-37-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0