1-Adamantyl isocyanate, 98%
1-Adamantyl isocyanate, 98%
1-Adamantyl isocyanate, 98%
1-Adamantyl isocyanate, 98%
Thermo Scientific Chemicals
1-Adamantyl isocyanate, 98%
CAS: 4411-25-0 | C11H15NO | 177.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA314360250
Price (MYR)
3,683.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,683.00
EA
Add to cart
Specifications
Chemical Name or Material1-Adamantyl isocyanate
CAS4411-25-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0