3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%
3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%
3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%
3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%
Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%
CAS: 16588-74-2 | C9H3F6NO | 255.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA314170050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 5-Bis(trifluoromethyl)phenyl isocyanate
Name Note98%
CAS16588-74-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning