Octadecyltrimethoxysilane, 90%, Tech.
Octadecyltrimethoxysilane, 90%, Tech.
Octadecyltrimethoxysilane, 90%, Tech.
Octadecyltrimethoxysilane, 90%, Tech.
Thermo Scientific Chemicals
Octadecyltrimethoxysilane, 90%, Tech.
CAS: 3069-42-9 | C21H46O3Si | 374.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA313601000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialOctadecyltrimethoxysilane
CAS3069-42-9
CAS Min %88.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0