3-Aminopropyltrimethoxysilane, 95%
3-Aminopropyltrimethoxysilane, 95%
3-Aminopropyltrimethoxysilane, 95%
3-Aminopropyltrimethoxysilane, 95%
Thermo Scientific Chemicals
3-Aminopropyltrimethoxysilane, 95%
CAS: 13822-56-5 | C6H17NO3Si | 179.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA313255000
Price (MYR)
1,222.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,222.00
EA
Add to cart
Specifications
Chemical Name or Material3-Aminopropyltrimethoxysilane
CAS13822-56-5
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0