alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%
alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%
alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%
alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%
Thermo Scientific Chemicals
alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%
CAS: 402-49-3 | C8H6BrF3 | 239.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA312990050
Price (MYR)
445.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
445.00
BOT
Add to cart
Specifications
Chemical Name or Materialα'-Bromo-α, α, α-trifluoro-p-xylene
Name Note98%
CAS402-49-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger