2,6-Difluorophenol, 98%
2,6-Difluorophenol, 98%
2,6-Difluorophenol, 98%
2,6-Difluorophenol, 98%
2,6-Difluorophenol, 98%
CAS: 28177-48-2 | C6H4F2O | 130.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA311630250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 6-Difluorophenol
CAS28177-48-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0