Octyl acetate, 99+%
Octyl acetate, 99+%
Octyl acetate, 99+%
Thermo Scientific Chemicals
Octyl acetate, 99+%
CAS: 112-14-1 | C10H20O2 | 172.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA310630050
Price (MYR)
187.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
187.00
BOT
Add to cart
Specifications
Chemical Name or MaterialOctyl acetate
Name Note99+%
CAS112-14-1
CAS Min %99.0
Health Hazard 1