Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
FSA310182500250 gGlass bottle
Catalog number FSA310182500
Price (MYR)
11,254.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
11,254.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium sulfide
Name Note90%, balance water
CAS1313-82-2
ColorBeige-Yellow
Health Hazard 1GHS Signal Word: Danger