4-(Methylthio)aniline, 98%, Thermo Scientific Chemicals
4-(Methylthio)aniline, 98%, Thermo Scientific Chemicals
4-(Methylthio)aniline, 98%, Thermo Scientific Chemicals
4-(Methylthio)aniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Methylthio)aniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA309830050
Price (MYR)
450.00
BOT
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
450.00
BOT
Add to cart
Specifications
Chemical Name or Material4-(Methylthio)aniline
Name Note98%
CAS104-96-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning