4-Methoxyphenylboronic acid, 97%
4-Methoxyphenylboronic acid, 97%
4-Methoxyphenylboronic acid, 97%
4-Methoxyphenylboronic acid, 97%
Thermo Scientific Chemicals
4-Methoxyphenylboronic acid, 97%
CAS: 5720-07-0 | C7H9BO3 | 151.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA309480050
Price (MYR)
570.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
570.00
EA
Add to cart
Specifications
Chemical Name or Material4-Methoxyphenylboronic acid
CAS5720-07-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0