Nonafluoropentanoic acid, 97%
Nonafluoropentanoic acid, 97%
Nonafluoropentanoic acid, 97%
Nonafluoropentanoic acid, 97%
Thermo Scientific Chemicals
Nonafluoropentanoic acid, 97%
CAS: 2706-90-3 | C5HF9O2 | 264.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA309290250
Price (MYR)
2,592.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,592.00
EA
Add to cart
Specifications
Chemical Name or MaterialNonafluoropentanoic acid
CAS2706-90-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0