Tetracontane, 98%
Tetracontane, 98%
Tetracontane, 98%
Thermo Scientific Chemicals
Tetracontane, 98%
CAS: 4181-95-7 | C40H82 | 563.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Catalog number FSA309165000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetracontane
CAS4181-95-7
Melting Point80.0°C to 83.0°C
ColorWhite
EINECS Number224-055-7