1-Bromo-3-methyl-2-butene, 96%
1-Bromo-3-methyl-2-butene, 96%
1-Bromo-3-methyl-2-butene, 96%
1-Bromo-3-methyl-2-butene, 96%
Thermo Scientific Chemicals
1-Bromo-3-methyl-2-butene, 96%
CAS: 870-63-3 | C5H9Br | 149.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number FSA307410100
Price (MYR)
562.00
BOT
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
562.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Bromo-3-methyl-2-butene
Name Note96%
CAS870-63-3
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger