2-Bromo-4'-nitroacetophenone, 95%
2-Bromo-4'-nitroacetophenone, 95%
2-Bromo-4'-nitroacetophenone, 95%
2-Bromo-4'-nitroacetophenone, 95%
Thermo Scientific Chemicals
2-Bromo-4'-nitroacetophenone, 95%
CAS: 99-81-0 | C8H6BrNO3 | 244.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA305480100
Price (MYR)
367.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
367.00
EA
Add to cart
Specifications
Chemical Name or Material2-Bromo-4′-nitroacetophenone
CAS99-81-0
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0