Iron(II) acetate, 95%
Iron(II) acetate, 95%
Iron(II) acetate, 95%
Iron(II) acetate, 95%
Thermo Scientific Chemicals
Iron(II) acetate, 95%
CAS: 3094-87-9 | C4H6FeO4 | 173.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA305372500
Price (MYR)
10,814.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
10,814.00
EA
Add to cart
Specifications
Chemical Name or MaterialIron(II) acetate
Melting Point190.0°C
Name Note95%
CAS3094-87-9
ColorBrown or Gray to White