Dodecanedioyl dichloride, 98%
Dodecanedioyl dichloride, 98%
Dodecanedioyl dichloride, 98%
Dodecanedioyl dichloride, 98%
Thermo Scientific Chemicals
Dodecanedioyl dichloride, 98%
CAS: 4834-98-4 | C12H20Cl2O2 | 267.2 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA30534025025 gGlass bottle
Catalog number FSA305340250
Price (MYR)
753.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
753.00
EA
Add to cart
Specifications
Chemical Name or MaterialDodecanedioyl dichloride
Name Note98%
CAS4834-98-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement