Methyl stearate, mixture of homologs
Methyl stearate, mixture of homologs
Methyl stearate, mixture of homologs
Methyl stearate, mixture of homologs
Thermo Scientific Chemicals
Methyl stearate, mixture of homologs
CAS: 112-61-8 | C19H38O2 | 298.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass Bottle
Catalog number FSA304220010
Price (MYR)
711.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
711.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl stearate
Acidity1mg KOH/g max.
CAS112-61-8
Melting Point37.0°C to 39.0°C
ColorCream to White or Yellow