3,5-Bis(trifluoromethyl)benzyl bromide, 97%
3,5-Bis(trifluoromethyl)benzyl bromide, 97%
3,5-Bis(trifluoromethyl)benzyl bromide, 97%
3,5-Bis(trifluoromethyl)benzyl bromide, 97%
Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)benzyl bromide, 97%
CAS: 32247-96-4 | C9H5BrF6 | 307.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA303640050
Price (MYR)
503.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
503.00
EA
Add to cart
Specifications
Chemical Name or Material3, 5-Bis(trifluoromethyl)benzyl bromide
CAS32247-96-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0