alpha-Bromo-2,6-difluorotoluene, 97%
alpha-Bromo-2,6-difluorotoluene, 97%
alpha-Bromo-2,6-difluorotoluene, 97%
alpha-Bromo-2,6-difluorotoluene, 97%
Thermo Scientific Chemicals
alpha-Bromo-2,6-difluorotoluene, 97%
CAS: 85118-00-9 | C7H5BrF2 | 207.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA303570050
Price (MYR)
325.00
G
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
325.00
G
Add to cart
Specifications
Chemical Name or Materialα-Bromo-2, 6-difluorotoluene
CAS85118-00-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0