2,5-Difluorobenzaldehyde, 98%
2,5-Difluorobenzaldehyde, 98%
2,5-Difluorobenzaldehyde, 98%
2,5-Difluorobenzaldehyde, 98%
Thermo Scientific Chemicals
2,5-Difluorobenzaldehyde, 98%
CAS: 2646-90-4 | C7H4F2O | 142.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA303520050
Price (MYR)
416.00
G
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
416.00
G
Add to cart
Specifications
Chemical Name or Material2, 5-Difluorobenzaldehyde
CAS2646-90-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0