4-Nitrophenyl-beta-D-glucopyranoside, 99%
4-Nitrophenyl-beta-D-glucopyranoside, 99%
4-Nitrophenyl-beta-D-glucopyranoside, 99%
4-Nitrophenyl-beta-D-glucopyranoside, 99%
Thermo Scientific Chemicals
4-Nitrophenyl-beta-D-glucopyranoside, 99%
CAS: 2492-87-7 | C12H15NO8 | 301.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA303110250
Price (MYR)
9,198.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
9,198.00
EA
Add to cart
Specifications
Chemical Name or Material4-Nitrophenyl-β-D-glucopyranoside
CAS2492-87-7
CAS Min %98.5
CAS Max %100.0
Melting Point165.0°C to 168.0°C