Scopoletin, 95%
Scopoletin, 95%
Scopoletin, 95%
Thermo Scientific Chemicals
Scopoletin, 95%
CAS: 92-61-5 | C10H8O4 | 192.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
50 mg
1 g
Packaging:
Glass bottle
Catalog number FSA302902500
Price (MYR)
1,708.00
EA
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,708.00
EA
Add to cart
Specifications
Chemical Name or MaterialScopoletin
Name Note95%
CAS92-61-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: