Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals
Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals
Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals
Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Plastic bottle
Catalog number FSA302865000
Price (MYR)
324.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
324.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydrogen peroxide
ColorColorless
CAS7722-84-1
Boiling Point114.0°C
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May intensify fire; oxidizer.
May cause respiratory irritation.
View more
Oxidation reagent
RUO – Research Use Only