3,4-(Methylenedioxy)phenylacetic acid, 98.5+%
3,4-(Methylenedioxy)phenylacetic acid, 98.5+%
3,4-(Methylenedioxy)phenylacetic acid, 98.5+%
3,4-(Methylenedioxy)phenylacetic acid, 98.5+%
Thermo Scientific Chemicals
3,4-(Methylenedioxy)phenylacetic acid, 98.5+%
CAS: 2861-28-1 | C9H8O4 | 180.159 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number FSA302620025
Price (MYR)
284.00
BOT
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
284.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 4-(Methylenedioxy)phenylacetic acid
CAS2861-28-1
CAS Min %98.5
CAS Max %100.0
Health Hazard 2GHS H Statement: