2-Chloroethylphosphonic acid, 90%, Tech.
2-Chloroethylphosphonic acid, 90%, Tech.
2-Chloroethylphosphonic acid, 90%, Tech.
2-Chloroethylphosphonic acid, 90%, Tech.
Thermo Scientific Chemicals
2-Chloroethylphosphonic acid, 90%, Tech.
CAS: 16672-87-0 | C2H6ClO3P | 144.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA301630050
Price (MYR)
2,981.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,981.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Chloroethylphosphonic acid
Name Notetech., 90%
CAS16672-87-0
CAS Min %88.0
Health Hazard 1GHS Signal Word: Danger