alpha-Terpineol, 97+%
alpha-Terpineol, 97+%
alpha-Terpineol, 97+%
alpha-Terpineol, 97+%
Thermo Scientific Chemicals
alpha-Terpineol, 97+%
CAS: 98-55-5 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
2.5 kg
Catalog number FSA301615000
Price (MYR)
469.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
469.00
BOT
Add to cart
Specifications
Chemical Name or Materialα-Terpineol
Name Note99%
CAS98-55-5
CAS Min %97.0
Health Hazard 1GHS Signal Word: Warning