1-Amino-1-cyclopropanecarboxylic acid, 99%
1-Amino-1-cyclopropanecarboxylic acid, 99%
1-Amino-1-cyclopropanecarboxylic acid, 99%
1-Amino-1-cyclopropanecarboxylic acid, 99%
Thermo Scientific Chemicals
1-Amino-1-cyclopropanecarboxylic acid, 99%
CAS: 22059-21-8 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA301422500
Price (MYR)
794.00
MG
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
794.00
MG
Add to cart
Specifications
Chemical Name or Material1-Amino-1-cyclopropanecarboxylic acid
Name Note99%
CAS22059-21-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning