Lithium tert-butoxide, 99%
Lithium tert-butoxide, 99%
Lithium tert-butoxide, 99%
Lithium tert-butoxide, 99%
Thermo Scientific Chemicals
Lithium tert-butoxide, 99%
CAS: 1907-33-1 | C4H9LiO | 80.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number FSA301225000
Price (MYR)
11,029.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
11,029.00
EA
Add to cart
Specifications
Chemical Name or MaterialLithium tert-butoxide
CAS1907-33-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Harmful if swallowed.
Reacts violently with water.
Health Hazard 3GHS P Statement
Keep cool.
Protect from sunlight.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if