4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent
Thermo Scientific Chemicals
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent
CAS: 5808-22-0 | C10H6O8S2 | 318.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA300600250
Price (MYR)
699.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
699.00
EA
Add to cart
Specifications
Chemical Name or Material4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate
CAS5808-22-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/