Aluminum, 99.5%, pieces (2-10 mm)
Aluminum, 99.5%, pieces (2-10 mm)
Aluminum, 99.5%, pieces (2-10 mm)
Aluminum, 99.5%, pieces (2-10 mm)
Thermo Scientific Chemicals
Aluminum, 99.5%, pieces (2-10 mm)
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number FSA300212500
Price (MYR)
583.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
583.00
EA
Add to cart
Specifications
Chemical Name or MaterialAluminum
ColorGray
Name Note99.5%
CAS7429-90-5
Melting Point660.0°C