1,3-Dioxolane, 99.5+%, pure, stabilized
1,3-Dioxolane, 99.5+%, pure, stabilized
1,3-Dioxolane, 99.5+%, pure, stabilized
1,3-Dioxolane, 99.5+%, pure, stabilized
Thermo Scientific Chemicals
1,3-Dioxolane, 99.5+%, pure, stabilized
CAS: 646-06-0 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA299480025
Price (MYR)
1,046.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,046.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 3-Dioxolane
Name NoteStabilized
CAS646-06-0
CAS Min %99.4
Health Hazard 1GHS Signal Word: Danger