Bis(triphenylphosphine)palladium(II) chloride, 98%
Bis(triphenylphosphine)palladium(II) chloride, 98%
Bis(triphenylphosphine)palladium(II) chloride, 98%
Bis(triphenylphosphine)palladium(II) chloride, 98%
Thermo Scientific Chemicals
Bis(triphenylphosphine)palladium(II) chloride, 98%
CAS: 13965-03-2 | C36H30Cl2P2Pd | 701.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA299251000
Price (MYR)
32,561.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
32,561.00
EA
Add to cart
Specifications
Chemical Name or MaterialBis(triphenylphosphine)palladium(II) chloride
CAS13965-03-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement
May cause long lasting harmful effects to aquatic life.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.