Methanol-d, for NMR, 99.0 atom % D
Methanol-d, for NMR, 99.0 atom % D
Methanol-d, for NMR, 99.0 atom % D
Methanol-d, for NMR, 99.0 atom % D
Thermo Scientific Chemicals
Methanol-d, for NMR, 99.0 atom % D
CAS: 1455-13-6 | CH4O | 33.048 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass Bottle
Catalog number FSA299131000
Price (MYR)
1,889.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,889.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethanol-d
Density0.8100g/mL
Deuterium (D)99.0% min.
Flash Point12°C
PackagingGlass Bottle
View more
For NMR
RUO – Research Use Only