4-(Trifluoromethoxy)benzylamine, 97%
4-(Trifluoromethoxy)benzylamine, 97%
4-(Trifluoromethoxy)benzylamine, 97%
4-(Trifluoromethoxy)benzylamine, 97%
Thermo Scientific Chemicals
4-(Trifluoromethoxy)benzylamine, 97%
CAS: 93919-56-3 | C8H8F3NO | 191.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA297700010
Price (MYR)
351.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
351.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethoxy)benzylamine
Name Note97%
CAS93919-56-3
CAS Min %96.0
CAS Max %100.0