L(-)-Lactic acid, lithium salt, 99%, pure
L(-)-Lactic acid, lithium salt, 99%, pure
L(-)-Lactic acid, lithium salt, 99%, pure
L(-)-Lactic acid, lithium salt, 99%, pure
Thermo Scientific Chemicals
L(-)-Lactic acid, lithium salt, 99%, pure
CAS: 27848-80-2 | C3H5O3Li | 96.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA297611000
Price (MYR)
1,160.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,160.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(-)-Lactic acid, lithium salt
Name Note99%
CAS27848-80-2
Health Hazard 1HYGROSCOPIC
Melting Point>75.0°C