Hydroxypropyl-beta-cyclodextrin, 97%
Hydroxypropyl-beta-cyclodextrin, 97%
Hydroxypropyl-beta-cyclodextrin, 97%
Hydroxypropyl-beta-cyclodextrin, 97%
Thermo Scientific Chemicals
Hydroxypropyl-beta-cyclodextrin, 97%
CAS: 128446-35-5 | C44H75O36 | 1180.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass Bottle
Catalog number FSA297561000
Price (MYR)
2,629.00
G
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,629.00
G
Add to cart
Specifications
Chemical Name or MaterialHydroxypropyl-β-cyclodextrin
CAS128446-35-5
Melting Point278.0°C
ColorWhite to Yellow
Infrared SpectrumAuthentic
View more
Chelation/Complexation reagent
RUO – Research Use Only