beta-Ionone, 96%, synthetic
beta-Ionone, 96%, synthetic
beta-Ionone, 96%, synthetic
beta-Ionone, 96%, synthetic
Thermo Scientific Chemicals
beta-Ionone, 96%, synthetic
CAS: 14901-07-6 | C13H20O | 192.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA297131000
Price (MYR)
440.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
440.00
EA
Add to cart
Specifications
Chemical Name or Materialβ-Ionone
Name Note96%
CAS14901-07-6
CAS Min %95.0
CAS Max %100.0