(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
Thermo Scientific Chemicals
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
CAS: 49805-30-3 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA297060010
Price (MYR)
366.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
366.00
EA
Add to cart
Specifications
Chemical Name or Material(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Name Note98%
CAS49805-30-3
Absorbance(c=1, CH2Cl2) (1cm cell), (c=1, CH2Cl2) (1cm cell), 2 max. at 400nm
CAS Min %97.5