Vancomycin hydrochloride, >900 microgram/mg
Vancomycin hydrochloride, >900 microgram/mg
Vancomycin hydrochloride, >900 microgram/mg
Vancomycin hydrochloride, >900 microgram/mg
Thermo Scientific Chemicals
Vancomycin hydrochloride, >900 microgram/mg
Vancomycin hydrochloride (>900 microgram/mg), CAS # 1404-93-9, is an antibiotic from Streptomyces orientalis, that shows antibacterial activity towards methicillin-resistant (β-lactam-resistant) Staphylococci bacterial strains.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass Bottle
Catalog number FSA296990050
Price (MYR)
5,107.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
5,107.00
EA
Add to cart
Specifications
Chemical Name or MaterialVancomycin hydrochloride
TypeVancomycin Hydrochloride
Physical FormPowder
CAS1404-93-9
pH2.5 to 4.5 (5% soln.)
View more
General Description
  • Vancomycin hydrochloride is a glycopeptide antibiotic and antiviral from Streptomyces orientalis
  • It shows antibiotic activity towards methicillin-resistant (β-lactam-resistant) Staphylococci bacterial strains
  • Vancomycin hydrochloride inhibits bacterial mucopeptide synthesis
Applications
  • Vancomycin hydrochloride demonstrates potent activity against gram-positive bacteria
  • It has been shown in studies to be highly effective against methicillin-resistant Staphylococcus aureus (MRSA) in vitro and in vivo
RUO – Research Use Only