Phenylhydrazine, 97%
Phenylhydrazine, 97%
Phenylhydrazine, 97%
Phenylhydrazine, 97%
Thermo Scientific Chemicals
Phenylhydrazine, 97%
CAS: 100-63-0 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA296685000
Price (MYR)
770.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
770.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenylhydrazine
CAS100-63-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Suspected of causing genetic defects.
May cause cancer.
Toxic if inhaled.
Toxic in contact with skin.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre