5-Hexyn-1-ol, 97%
5-Hexyn-1-ol, 97%
5-Hexyn-1-ol, 97%
Thermo Scientific Chemicals
5-Hexyn-1-ol, 97%
CAS: 928-90-5 | C6H10O | 98.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA296580050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material5-Hexyn-1-ol
CAS928-90-5
Health Hazard 1GHS Signal Word: Warning
ColorColorless to Yellow
Density0.8900g/mL