Rhodamine B, 98+%, pure
Rhodamine B, 98+%, pure
Rhodamine B, 98+%, pure
Rhodamine B, 98+%, pure
Thermo Scientific Chemicals
Rhodamine B, 98+%, pure
CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA296571000
Price (MYR)
401.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
401.00
EA
Add to cart
Specifications
Chemical Name or MaterialRhodamine B
Melting Point210.0°C to 211.0°C
Name NotePure
CAS81-88-9
Health Hazard 1Danger