2,2,3,3-Tetrafluoro-1-propanol, 99+%
2,2,3,3-Tetrafluoro-1-propanol, 99+%
2,2,3,3-Tetrafluoro-1-propanol, 99+%
2,2,3,3-Tetrafluoro-1-propanol, 99+%
Thermo Scientific Chemicals
2,2,3,3-Tetrafluoro-1-propanol, 99+%
CAS: 76-37-9 | C3H4F4O | 132.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA296381000
Price (MYR)
2,494.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,494.00
EA
Add to cart
Specifications
Chemical Name or Material2,2,3,3-Tetrafluoro-1-propanol
CAS76-37-9
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0