(Chloromethylene)dimethylammonium chloride, 96%
(Chloromethylene)dimethylammonium chloride, 96%
(Chloromethylene)dimethylammonium chloride, 96%
(Chloromethylene)dimethylammonium chloride, 96%
Thermo Scientific Chemicals
(Chloromethylene)dimethylammonium chloride, 96%
CAS: 3724-43-4 | C3H7Cl2N | 128 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA295770050
Price (MYR)
373.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
373.00
BOT
Add to cart
Specifications
Chemical Name or Material(Chloromethylene)dimethylammonium chloride
Name Note96%
CAS3724-43-4
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger
View more
Alkylation reagent
RUO – Research Use Only