Ethylene glycol, 99.5%, for analysis
Ethylene glycol, 99.5%, for analysis
Ethylene glycol, 99.5%, for analysis
Ethylene glycol, 99.5%, for analysis
Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number FSA295530250
Price (MYR)
4,437.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
4,437.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthylene glycol
Melting Point-13.0°C
CAS107-21-1
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
Harmful if swallowed.
View more
For analysis
RUO – Research Use Only