Chromosulfuric acid, Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA295510010
Price (MYR)
606.00
LT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
606.00
LT
Add to cart
Specifications
Chemical Name or MaterialChromosulfuric acid
ColorBrown to Brown-Red
CAS65272-71-1
Boiling Point330.0°C
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only