2-Chloropropionyl chloride, 95%
2-Chloropropionyl chloride, 95%
2-Chloropropionyl chloride, 95%
2-Chloropropionyl chloride, 95%
Thermo Scientific Chemicals
2-Chloropropionyl chloride, 95%
CAS: 7623-09-8 | C3H4Cl2O | 126.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA295160250
Price (MYR)
389.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
389.00
EA
Add to cart
Specifications
Chemical Name or Material2-Chloropropionyl chloride
Name Note95%
CAS7623-09-8
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger