2,3-Naphthalenedicarboxaldehyde, 95%
2,3-Naphthalenedicarboxaldehyde, 95%
2,3-Naphthalenedicarboxaldehyde, 95%
2,3-Naphthalenedicarboxaldehyde, 95%
Thermo Scientific Chemicals
2,3-Naphthalenedicarboxaldehyde, 95%
CAS: 7149-49-7 | C12H8O2 | 184.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass Bottle
Catalog number FSA294721000
Price (MYR)
1,047.00
EA
Add to cart
Quantity:
100 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,047.00
EA
Add to cart
Specifications
Chemical Name or Material2, 3-Naphthalenedicarboxaldehyde
CAS7149-49-7
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0